missing translation for 'onlineSavingsMsg'
Learn More

Benzonitrile, Reagent Grade, 99%, Honeywell™

Product Code. p-7120220 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
100 mL
1 L
Packaging:
Glass Bottle
Unit Size:
1 L
100mL
This item is not returnable. View return policy

Product Code. 15697700

Brand: Honeywell Research Chemicals B8959100ML

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-47-0
Molecular Formula C7H5N
Molecular Weight (g/mol) 103.124
MDL Number MFCD00001770
InChI Key JFDZBHWFFUWGJE-UHFFFAOYSA-N
Synonym phenyl cyanide, cyanobenzene, benzenenitrile, benzoic acid nitrile, benzene, cyano, benzenecarbonitrile, phenylcyanide, fenylkyanid, fenylkyanid czech, benzonitril
PubChem CID 7505
ChEBI CHEBI:27991
IUPAC Name benzonitrile
SMILES C1=CC=C(C=C1)C#N

Specifications

Melting Point -13°C
Color Colorless
Boiling Point 191°C
Flash Point 70°C (158°F)
Packaging Glass Bottle
Linear Formula C6H5CN
Quantity 100 mL
UN Number UN2224
Beilstein 506893
Vapor Density 3.56 (Air = 1.0)
Formula Weight 103.12g/mol
Vapor Pressure 1 hPa at 20°C
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Benzonitrile
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.