missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NucA nuclease Antibody, Clone: 1028937, R&D Systems™

Mouse Monoclonal Antibody
£169.00 - £408.00
Specifications
| Antigen | NucA Nuclease |
|---|---|
| Clone | 1028937 |
| Applications | ELISA |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18794577
![]() |
R&D Systems
MAB100635-SP |
25 μg |
£169.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18701607
![]() |
R&D Systems
MAB100635-100 |
100 μg |
£408.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NucA Nuclease Monoclonal antibody specifically detects NucA Nuclease in samples. It is validated for ELISASpecifications
| NucA Nuclease | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| DcD | |
| E. coli-derived Benzonase Nuclease/NucA, Met1-Asn266, Accession # P13717 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1028937 | |
| Monoclonal | |
| Purified | |
| RUO | |
| P13717 | |
| 233874471 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title