missing translation for 'onlineSavingsMsg'
Learn More

Benzoisothiazol-3-one, TRC

Product Code. 17962465
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17962465 1 g 1g
30133240 10 g 10g
30012695 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17962465 Supplier Toronto Research Chemicals Supplier No. TRCB2040351G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2634-33-5
Molecular Formula C7 H5 N O S
Molecular Weight (g/mol) 151.19
Synonym 1,2-Benzisothiazolin-3-one (6CI,7CI,8CI), 1,2-Benzisothiazol-3-one, 1,2-Benzisothiazolone, 1,2-Benzoisothiazol-3-one, 2,3-Dihydrobenzisothiazol-3-one, 3-Hydroxy-1,2-benzisothiazole, AQ, AQ (antibacterial), Acticide B 20N, Acticide BIT, Acticide BW 20, Apizas AP-DS, BIT, BIT 10W, BIT 20, Benzisothiazolin-3-one, Benzisothiazolinone, Benzisothiazolone, Benzo[d]isothiazol-3(2H)-one, Benzocil, Benzoisothiazol-3-one, Bestcide 200K, Bioban BIT 20DPG, Bioban Ultra Bit, Canguard BIT, Canguard BIT 20AS-E, Canguard BIT 20DPG, Canguard Ultra BIT 20LE, Denicide BIT, Denicide BIT 20N, GXL, Koralone B 119, Mergal 753, Nipacide BIT, Nipacide BIT 10W, Nipacide BIT 20, Nuosept 485, Nuosept 491, Nuosept 495, Parmetol B 70, Parmetol D 11, Preventol BIT 20D, Proxel AB, Proxel AQ, Proxel BD, Proxel BD 20, Proxel BDN, Proxel CF, Proxel GXL, Proxel GXL(S), Proxel HL 2, Proxel LV, Proxel LV-S, Proxel PL, Proxel Press Paste, Proxel Press Paste D, Proxel TN, Proxel Ultra 5, Proxel XL, Proxel XL 2, Rocima 640, SD 202, SD 202 (bactericide), San-aibac AP, Topcide 600, Troysan 1050, XBINX
IUPAC Name 1,2-benzothiazol-3-one
SMILES O=C1NSc2ccccc12

Specifications

Analyte or Component Names 1,2-Benzisothiazol-3(2H)-one
Shipping Condition Room Temperature
InChI Formula InChI=1S/C7H5NOS/c9-7-5-3-1-2-4-6(5)10-8-7/h1-4H,(H,8,9)
Solution Type Neat
Industry Type Forensics
Quantity 1 g
Formula Weight 151.0092
Chemical Name or Material 1,2-Benzisothiazol-3(2H)-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.