missing translation for 'onlineSavingsMsg'
Learn More

Benzoin, TRC

Product Code. 30338769
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30338769 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30338769 Supplier Toronto Research Chemicals Supplier No. TRCB2040051G
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzoin
CAS 119-53-9
Molecular Formula C14 H12 O2
Quantity 1 g
Synonym Ethanone, 2-hydroxy-1,2-diphenyl-, Benzoin (8CI), 2-Hydroxy-1,2-diphenylethanone, (RS)-Benzoin, (±)-2-Hydroxy-1,2-diphenylethanone, (±)-2-Hydroxy-2-phenylacetophenone, (±)-Benzoin, 1,2-Diphenyl-1-hydroxy-2-ethanone, 1,2-Diphenyl-2-hydroxyethanone, 1,2-Diphenyl-2-oxoethanol, 2-Hydroxy-1,2-diphenyl-1-ethanone, 2-Hydroxy-1-phenyl-2-phenylethanone, 2-Hydroxy-2-phenylacetophenone, 2-Hydroxyl-1,2-phenylethanone, 2-Phenyl-2-hydroxyacetophenone, AW 63, Benzoylphenylcarbinol, DL-Benzoin, Desyl alcohol, Esacure EB 3, F 297, GK 19, Jiqi GK 19, L 307, LC Benzoin, NSC 8082, Nisso Cure MB, S 19, S 19 (benzoin), Seikuol Z, Wy 42956, dl-Benzoin, α-Benzoylbenzenemethanol, α-Hydroxy-α-phenylacetophenone, α-Hydroxybenzyl phenyl ketone
InChI Formula InChI=1S/C14H12O2/c15-13(11-7-3-1-4-8-11)14(16)12-9-5-2-6-10-12/h1-10,13,15H
SMILES OC(C(=O)c1ccccc1)c2ccccc2
IUPAC Name 2-hydroxy-1,2-diphenylethanone
Molecular Weight (g/mol) 212.24
Formula Weight 212.0837
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.