missing translation for 'onlineSavingsMsg'
Learn More

Benzoin methyl ether, 97%

Product Code. 11414043
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11414043 100 g 100g
11404043 25 g 25g
11424043 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11414043 Supplier Thermo Scientific Alfa Aesar Supplier No. A10296.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3524-62-7 | C15H14O2 | 226.275 g/mol

Benzoin methyl ether is used in UV curing inks, wood coatings, paper coatings, optical fiber, PCB, screen printing , paper varnish and other surface coatings. It is used for anticratering in powder coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzoin methyl ether is used in UV curing inks, wood coatings, paper coatings, optical fiber, PCB, screen printing , paper varnish and other surface coatings. It is used for anticratering in powder coatings.

Solubility
Soluble in methanol.

Notes
Store away from oxidizing agents. Store in a cool, dry condition in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3524-62-7
Molecular Formula C15H14O2
Molecular Weight (g/mol) 226.275
MDL Number MFCD00008492
InChI Key BQZJOQXSCSZQPS-UHFFFAOYSA-N
Synonym benzoin methyl ether, methyl benzoin, benzoin methyl ester, o-methylbenzoin, 2-methoxy-2-phenylacetophenone, nisso cure mbo, ethanone, 2-methoxy-1,2-diphenyl, acetophenone, 2-methoxy-2-phenyl, qcu 3, alpha-methoxydeoxybenzoin
PubChem CID 98097
IUPAC Name 2-methoxy-1,2-diphenylethanone
SMILES COC(C1=CC=CC=C1)C(=O)C2=CC=CC=C2

Specifications

Melting Point 46°C to 50°C
Density 1.128
Boiling Point 188°C to 189°C (15 mmHg)
Flash Point >110°C (230°F)
Quantity 100 g
Beilstein 2052112
Merck Index 14,1093
Solubility Information Soluble in methanol.
Formula Weight 226.28
Percent Purity 97%
Chemical Name or Material Benzoin methyl ether
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.