missing translation for 'onlineSavingsMsg'
Learn More

Benzoic anhydride, 98%

Product Code. 11994971 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11994971 2.5 kg Plastic bottle 2.5kg
10697432 25 g Plastic bottle 25g
10297493 100 g Plastic bottle 100g
10204660 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11994971 Supplier Thermo Scientific Acros Supplier No. 105500025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 93-97-0 | C14H10O3 | 226.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-97-0
Molecular Formula C14H10O3
Molecular Weight (g/mol) 226.23
MDL Number MFCD00003073
InChI Key CHIHQLCVLOXUJW-UHFFFAOYSA-N
Synonym benzoic anhydride, benzoic acid, anhydride, benzoyl anhydride, benzoylbenzoate, benzoic acid anhydride, benzoic acid, 1,1'-anhydride, phenyl anhydride, benzoesaeureanhydrid, unii-9k7x34fov2, phenylcarbonyl benzoate
PubChem CID 7167
ChEBI CHEBI:38815
IUPAC Name benzoyl benzoate
SMILES C1=CC=C(C=C1)C(=O)OC(=O)C2=CC=CC=C2

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 38.0°C to 44.0°C
Color White
Density 1.1900g/mL
Boiling Point 360.0°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Plastic bottle
Linear Formula (C6H5CO)2O
Quantity 2.5 kg
Beilstein 09,164
Fieser 01,49; 05,23
Merck Index 15,1094
Specific Gravity 1.19
Solubility Information Solubility in water: 0,01g/L. Other solubilities: soluble in alcohol,chloroform,acetone,benzene,,ethyl acetate,toluene,xylene,ether,glacial,acetic acid,acetic anhydride,moderately soluble in petroleum ether
Formula Weight 226.23
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Benzoic anhydride, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Benzoic anhydride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.