missing translation for 'onlineSavingsMsg'
Learn More

Benzoic Acid, LoGiCal

Product Code. 30457852
Change view
Click to view available options
Quantity:
10 mg
Dimensione della confezione:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity unitSize
30457852 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Product Code. 30457852 Supplier LoGiCal Supplier No. LGCFOR0278.01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzoic acid
CAS 65-85-0
Analyte or Component Names Benzoic acid
Shipping Condition Room Temperature
Molecular Formula C7 H6 O2
Certifications/Compliance ISO 9001
Synonym Benfluorex Hydrochloride Imp. C (EP), Benfluorex Imp. C (EP), Glycopyrronium Bromide Imp. D (EP), Mefenamic Acid Imp. D (EP), Metronidazole Benzoate Imp. C (EP), Benzoic Acid, Benzenecarboxylic acid, Benzeneformic acid, Benzenemethanoic acid, Carboxybenzene, Dracylic acid, E 210, HA 1, HA 1 (acid), MENNO-Florades, NSC 149, Phenylcarboxylic acid, Phenylformic acid, Purox B, Retarder BA, Retardex, Salvo liquid, Solvo powder, Tenn-Plas, VevoVitall, BRL-9421A, Benfluorex Hydrochloride Impurity C, Benzoyl Peroxide Hydrous Impurity B, Glycopyrronium Bromide Impurity D, Mefenamic Acid Impurity D, Metronidazole Benzoate Impurity C, Benfluorex Impurity C, Glycopyrronium Impurity D
InChI Formula InChI=1S/C7H6O2/c8-7(9)6-4-2-1-3-5-6/h1-5H,(H,8,9)
Solution Type Neat
SMILES OC(=O)c1ccccc1
IUPAC Name benzoic acid
Industry Type Clinical Testing
Molecular Weight (g/mol) 122.12
Quantity 10 mg
Formula Weight 122.0368 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.