missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzoic Acid-d5, TRC
Specifications
Specifications
| Chemical Name or Material | Benzoic acid-d5 |
| CAS | 1079-02-3 |
| Molecular Formula | C7 2H5 H O2 |
| Quantity | 100 mg |
| Synonym | Benzoic-2,3,4,5,6-d5 acid, Benzoic-d5 acid (6CI,7CI,8CI,9CI), 2,3,4,5,6-Pentadeuteriobenzoic acid, Benzoic acid-d5, Pentadeuteriobenzoic acid, Benzoic Acid D5 (Phenyl D5), Benzoic acid D5 (phenyl D5) |
| InChI Formula | InChI=1S/C7H6O2/c8-7(9)6-4-2-1-3-5-6/h1-5H,(H,8,9)/i1D,2D,3D,4D,5D |
| SMILES | [2H]c1c([2H])c([2H])c(C(=O)O)c([2H])c1[2H] |
| IUPAC Name | 2,3,4,5,6-pentadeuteriobenzoic acid |
| Molecular Weight (g/mol) | 127.15 |
| Formula Weight | 127.0682 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?