Learn More
Benzoic acid, 99%, Thermo Scientific Chemicals
Use promo code "21741" to get your promotional price.
Description
Benzoic acid is an important precursor for the preparation of many other organic substances viz. benzoyl chloride, benylbenzoate, phenol, sodium benzoate, benzyl alcohol as well as benzoate plasticizers such as glycol-, diethhyleneglycol- and triethyleneglycol esters. Benzoic acid and its salts are used as food preservatives. It is involved in rubber polymerization as an activator and a retardant. It is the main component of benzoin resin and is a constituent of Whitfield’s ointment, which is used for the treatment of fungal skin diseases such as tinea, ringworm and athlete’s foot. It is widely used in cosmetics, dyes, plastics and insect repellents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Benzoic acid is an important precursor for the preparation of many other organic substances viz. benzoyl chloride, benylbenzoate, phenol, sodium benzoate, benzyl alcohol as well as benzoate plasticizers such as glycol-, diethhyleneglycol- and triethyleneglycol esters. Benzoic acid and its salts are used as food preservatives. It is involved in rubber polymerization as an activator and a retardant. It is the main component of benzoin resin and is a constituent of Whitfield’s ointment, which is used for the treatment of fungal skin diseases such as tinea, ringworm and athlete’s foot. It is widely used in cosmetics, dyes, plastics and insect repellents.
Solubility
Soluble in acetone, benzene, chloroform, alcohol, diethyl ether, hexane and liquid ammonia. Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents, reducing agents and strong bases.
Specifications
Melting Point | 121°C to 124°C |
Boiling Point | 249°C |
Assay Percent Range | 99% |
Beilstein | 636131 |
Merck Index | 14,1091 |
Solubility Information | Soluble in acetone,benzene,chloroform,alcohol,diethyl ether,hexane and liquid ammonia. Slightly soluble in water. |
Formula Weight | 122.12 |
Percent Purity | 99% |
Density | 1.32 |
Quantity | 250 g |
Show More |
Safety and Handling
- Benzoic acid
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 1
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H372-Causes damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : benzoic acid
RUO – Research Use Only