missing translation for 'onlineSavingsMsg'
Learn More

Benzoic acid, 99.6%, ACS reagent, Thermo Scientific Chemicals

Product Code. 10799911
Click to view available options
Quantity:
25 g
500 g
2 kg
10 kg
Packaging:
Plastic bottle
Plastic drum

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65-85-0
Molecular Formula C7H6O2
Molecular Weight (g/mol) 122.12
InChI Key WPYMKLBDIGXBTP-UHFFFAOYSA-N
Synonym benzenecarboxylic acid, dracylic acid, benzeneformic acid, carboxybenzene, phenylformic acid, benzenemethanoic acid, phenylcarboxylic acid, retardex, benzoesaeure gk, benzoesaeure gv
PubChem CID 243
ChEBI CHEBI:30746
IUPAC Name benzoic acid
SMILES C1=CC=C(C=C1)C(=O)O

Specifications

CAS Min % 99.5
CAS Max % 100.0
Melting Point 121.0°C to 123.0°C
Color White
Boiling Point 249.0°C
Flash Point 121°C
Ignition Residue 0.005% max.
Infrared Spectrum Authentic
Assay Percent Range 99.6%
Packaging Plastic bottle
Linear Formula C6H5CO2H
Quantity 2 kg
Beilstein 09,92
Fieser 01,49; 13,312
Merck Index 15,1093
Solubility Information Solubility in water: 3g/L (20°C). Other solubilities: soluble in alcohol,benzene,ether,chloroform,,ccl4,carbon disulfide and turpentine
Formula Weight 122.12
Percent Purity 99.6%
Grade ACS Reagent
Insoluble Matter 0.005% max. (in methanol)
Physical Form Crystalline Powder
Chemical Name or Material Benzoic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'