missing translation for 'onlineSavingsMsg'
Learn More

Benzoic acid, 99.5%, for analysis

Product Code. 10289773 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10289773 5 kg Plastic drum 5kg
10432801 250 g Plastic bottle 250g
10422801 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10289773 Supplier Thermo Scientific Acros Supplier No. 221800050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 65-85-0 | C7H6O2 | 122.123 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65-85-0
Molecular Formula C7H6O2
Molecular Weight (g/mol) 122.123
MDL Number MFCD00002398
InChI Key WPYMKLBDIGXBTP-UHFFFAOYSA-N
Synonym benzenecarboxylic acid, dracylic acid, benzeneformic acid, carboxybenzene, phenylformic acid, benzenemethanoic acid, phenylcarboxylic acid, retardex, benzoesaeure gk, benzoesaeure gv
PubChem CID 243
ChEBI CHEBI:30746
IUPAC Name benzoic acid
SMILES C1=CC=C(C=C1)C(=O)O

Specifications

Melting Point 121.0°C to 123.0°C
Boiling Point 249.0°C
CAS Min % 99.4
CAS Max % 100.0
Color White
Assay Percent Range 99.5%
Linear Formula C6H5CO2H
Beilstein 09, 92
Fieser 01,49; 13,312
Merck Index 15, 1093
Solubility Information Solubility in water: 3g/L (20°C). Other solubilities: soluble in alcohol, benzene, ether, chloroform,, ccl4, carbon disulfide and turpentine
Formula Weight 122.12
Percent Purity 99.5%
Grade Analytical
Density 121g/mL
Quantity 5 kg
Flash Point 121°C
Infrared Spectrum Authentic
Packaging Plastic drum
Physical Form Fine Crystalline Powder
Chemical Name or Material Benzoic acid, For analysis
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.