missing translation for 'onlineSavingsMsg'
Learn More

Benzoguanamine, 99%

Product Code. 11394589
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11394589 100 g 100g
11410711 500 g 500g
11420711 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11394589 Supplier Thermo Scientific Alfa Aesar Supplier No. B20157.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 91-76-9 | C9H9N5 | 187.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-76-9
Molecular Formula C9H9N5
Molecular Weight (g/mol) 187.21
MDL Number MFCD00023187
InChI Key GZVHEAJQGPRDLQ-UHFFFAOYSA-N
Synonym benzoguanamine, 2,4-diamino-6-phenyl-1,3,5-triazine, benzoguanimine, 1,3,5-triazine-2,4-diamine, 6-phenyl, usaf rh-5, 2,4-diamino-6-phenyl-s-triazine, 2-phenyl-4,6-diamino-s-triazine, 4,6-diamino-2-phenyl-s-triazine, benzoguanamine van, s-triazine, 2,4-diamino-6-phenyl
PubChem CID 7064
IUPAC Name 6-phenyl-1,3,5-triazine-2,4-diamine
SMILES NC1=NC(=NC(N)=N1)C1=CC=CC=C1

Specifications

Melting Point 225°C to 228°C
Quantity 100 g
Beilstein 153223
Merck Index 14,1089
Formula Weight 187.21
Percent Purity 99%
Chemical Name or Material Benzoguanamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.