missing translation for 'onlineSavingsMsg'
Learn More

Benzocaine, TRC

Product Code. 30132624
Change view
Click to view available options
Quantity:
5 g
100 g
Unit Size:
100g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132624 100 g 100g
30041068 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30132624 Supplier Toronto Research Chemicals Supplier No. TRCB202970100G
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-09-7
Molecular Formula C9 H11 N O2
Molecular Weight (g/mol) 165.19
Synonym Benzoic acid, p-amino-, ethyl ester (8CI), (p-(Ethoxycarbonyl)phenylamine, 4-(Ethoxycarbonyl)aniline, 4-(Ethoxycarbonyl)phenylamine, 4-Aminobenzoic acid ethyl ester, 4-Carbethoxyaniline, Aethoform, Amben ethyl ester, Americaine, Anaesthan-syngala, Anaesthesin, Anaesthin, Anbesol, Anestezin, Anesthesin, Anesthesine, Anesthone, Baby Anbesol, Benzoak, Benzocaine, Diet Ayds, Et-PABA, Ethoform, Ethyl 4-aminobenzoate, Ethyl aminobenzoate, Ethyl p-aminobenzenecarboxylate, Ethyl p-aminobenzoate, Ethyl p-aminophenylcarboxylate, Gingicaine, Identhesin, Keloform, NSC 41531, NSC 4688, Norcain, Norcaine, Ora-jel, Orabase-B, Orthesin, Parathesin, Parathesine, Slim Mint Gum, Solu H, p-(Ethoxycarbonyl)aniline, p-Aminobenzoic acid ethyl ester, p-Carbethoxyaniline, p-Ethoxycarboxylic aniline
IUPAC Name ethyl 4-aminobenzoate
SMILES CCOC(=O)c1ccc(N)cc1

Specifications

InChI Formula InChI=1S/C9H11NO2/c1-2-12-9(11)7-3-5-8(10)6-4-7/h3-6H,2,10H2,1H3
Formula Weight 165.079
Percent Purity >95%
Quantity 100 g
Chemical Name or Material 4-Aminobenzoic acid ethyl ester (Benzocaine)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.