missing translation for 'onlineSavingsMsg'
Learn More

Benzo[g]chrysene, 98%, Thermo Scientific™

Product Code. 15572383 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15572383 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15572383 Supplier Thermo Scientific Acros Supplier No. 460270010
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Spécification

CAS 196-78-1
Melting Point 126.0°C
Boiling Point 525.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Molecular Formula C22H14
Quantity 1 g
Synonym benzo g chrysene, benzo a triphenylene, naphthophenanthrene, unii-e7mq64xu75, 1,2,3,4-dibenzphenanthrene, 1,2,3,4-dibenzophenanthrene, 1,2:3,4:7,8-tribenznaphthalene, benzochrysene, acmc-20gx0y
InChI Key JZOIZKBKSZMVRV-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C=CC3=C2C4=CC=CC=C4C5=CC=CC=C35
Molecular Weight (g/mol) 278.35
PubChem CID 9140
Formula Weight 278.35
Percent Purity 98%
Chemical Name or Material Benzo[g]chrysene
Afficher plus Afficher moins
compliance-icons
Identifiant produit
  • Benzo[g]chrysene
Mention d'avertissement
  • Warning
Catégorie de danger
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Mention de danger
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Conseils de prudence
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.