missing translation for 'onlineSavingsMsg'
Learn More

Benzo[c]cinnoline, 99%, Thermo Scientific™

Product Code. 10326120 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 10326120

Brand: Thermo Scientific Acros 105460010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 230-17-1
CAS Min % 98.5
CAS Max % 100.0
Melting Point 156.0°C to 159.0°C
Color Yellow to Green
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C12H8N2
MDL Number MFCD00004990
Quantity 1 g
Synonym benzo c cinnoline, diphenylenazone, 2,2'-azobiphenyl, 5,6-phenanthroline, 9,10-diazaphenanthrene, 3,4-benzocinnoline, 5,6-diazaphenanthrene, o-diphenyleneazine, phenazone three-ring system
InChI Key SWJXWSAKHXBQSY-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3N=N2
IUPAC Name benzo[c]cinnoline
Molecular Weight (g/mol) 180.21
PubChem CID 9190
Formula Weight 180.21
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Benzo[c]cinnoline, 99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.