missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]thiophene, 98+%

Product Code. 11479443
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11479443 100 g 100g
11459443 5 g 5g
11469443 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11479443 Supplier Thermo Scientific Alfa Aesar Supplier No. A12328.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 95-15-8 | C8H6S | 134.196 g/mol

Benzo[b]thiophene is used in the preparation of 2-thianapthenylphenyllithium, via metalation by n-butyllithium. It is used in organic as well as pharmaceutical industry as a component in the synthesis of raloxifene, a breast cancer therapeutic, or hydrodesulfurization reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzo[b]thiophene is used in the preparation of 2-thianapthenylphenyllithium, via metalation by n-butyllithium. It is used in organic as well as pharmaceutical industry as a component in the synthesis of raloxifene, a breast cancer therapeutic, or hydrodesulfurization reactions.

Solubility
Soluble in water (0.13 g/L at 20°C), chloroform and methanol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-15-8
Molecular Formula C8H6S
Molecular Weight (g/mol) 134.196
MDL Number MFCD00005864
InChI Key FCEHBMOGCRZNNI-UHFFFAOYSA-N
Synonym benzo b thiophene, thianaphthene, benzothiophene, benzothiofuran, thionaphthene, 1-thiaindene, thianaphthen, benzothiophen, thianaphtene, 1-benzothiophen
PubChem CID 7221
ChEBI CHEBI:35858
IUPAC Name 1-benzothiophene
SMILES C1=CC=C2C(=C1)C=CS2

Specifications

Melting Point 28°C to 32°C
Density 1.149
Boiling Point 221°C to 222°C
Flash Point 91°C (195°F)
Odor Unpleasant
Quantity 100 g
UN Number UN3077
Beilstein 80580
Merck Index 14,9303
Solubility Information Soluble in water (0.13g/L at 20°C),chloroform and methanol.
Formula Weight 134.2
Percent Purity ≥98%
Physical Form Crystals or Powder or Crystalline Powder or Fused Solid
Chemical Name or Material Benzo[b]thiophene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.