missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzo[b]thiophene, 98+%, Thermo Scientific Chemicals
15.93 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Benzo[b]thiophene is used in the preparation of 2-thianapthenylphenyllithium, via metalation by n-butyllithium. It is used in organic as well as pharmaceutical industry as a component in the synthesis of raloxifene, a breast cancer therapeutic, or hydrodesulfurization reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Benzo[b]thiophene is used in the preparation of 2-thianapthenylphenyllithium, via metalation by n-butyllithium. It is used in organic as well as pharmaceutical industry as a component in the synthesis of raloxifene, a breast cancer therapeutic, or hydrodesulfurization reactions.
Solubility
Soluble in water (0.13 g/L at 20°C), chloroform and methanol.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Benzo[b]thiophene is used in the preparation of 2-thianapthenylphenyllithium, via metalation by n-butyllithium. It is used in organic as well as pharmaceutical industry as a component in the synthesis of raloxifene, a breast cancer therapeutic, or hydrodesulfurization reactions.
Solubility
Soluble in water (0.13 g/L at 20°C), chloroform and methanol.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Melting Point | 28°C to 32°C |
Density | 1.149 |
Boiling Point | 221°C to 222°C |
Flash Point | 91°C (195°F) |
Odor | Unpleasant |
Quantity | 5 g |
UN Number | UN3077 |
Beilstein | 80580 |
Merck Index | 14,9303 |
Solubility Information | Soluble in water (0.13g/L at 20°C),chloroform and methanol. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'