missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]thiophene-3-sulfonyl chloride, 97%

Product Code. 15427898
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15427898

Brand: Thermo Scientific Alfa Aesar H61368.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18494-87-6 | C8H5ClO2S2 | 232.696 g/mol

Benzo[b]thiophene-3-sulfonyl chloride is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzo[b]thiophene-3-sulfonyl chloride is used as a pharmaceutical intermediate.

Solubility
Sparingly soluble in water (0.091 g/L at 25°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18494-87-6
Molecular Formula C8H5ClO2S2
Molecular Weight (g/mol) 232.696
MDL Number MFCD02681897
InChI Key WYYRQMZTUXFUTC-UHFFFAOYSA-N
Synonym benzothiophene-3-sulfonyl chloride, benzo b thiophene-3-sulfonyl chloride, benzo b thiophene-3-sulfonylchloride, benzo b thiophene-3-sulphonyl chloride, pubchem14540, 3-chlorosulphonyl-benzothiophene, benzothiophene-3-sulfonylchloride, 3-chlorosulphonyl-1-benzothiophene, benzo b thiophen-3-ylchlorosulfone
PubChem CID 2776340
IUPAC Name 1-benzothiophene-3-sulfonyl chloride
SMILES C1=CC=C2C(=C1)C(=CS2)S(=O)(=O)Cl

Specifications

Melting Point 86°C to 88°C
Quantity 1 g
UN Number UN3261
Beilstein 1285265
Sensitivity Moisture sensitive
Solubility Information Sparingly soluble in water (0.091g/L at 25°C).
Formula Weight 232.71
Percent Purity 97%
Chemical Name or Material Benzo[b]thiophene-3-sulfonyl chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.