missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]thiophene-3-carboxylic acid, 96%

Product Code. 15467857
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15467857 25 g 25g
11474540 1 g 1g
11484540 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15467857 Supplier Thermo Scientific Alfa Aesar Supplier No. H50105.14
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5381-25-9 | C9H6O2S | 178.205 g/mol

Benzo[b]thiophene-3-carboxylic acid is used as organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzo[b]thiophene-3-carboxylic acid is used as organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5381-25-9
Molecular Formula C9H6O2S
Molecular Weight (g/mol) 178.205
MDL Number MFCD01846406
InChI Key DRBLTQNCQJXSNU-UHFFFAOYSA-N
Synonym benzo b thiophene-3-carboxylic acid, benzothiophene-3-carboxylic acid, benzothiophene-3-carboxylicacid, pubchem13463, acmc-20a10z, ksc178m8d, 1-benzo b thiophene-3-carboxylic acid, #, thianaphthene-3-carboxylic acid
PubChem CID 601280
IUPAC Name 1-benzothiophene-3-carboxylic acid
SMILES C1=CC=C2C(=C1)C(=CS2)C(=O)O

Specifications

Melting Point 174°C to 176°C
Quantity 25 g
Formula Weight 178.22
Percent Purity 96%
Chemical Name or Material Benzo[b]thiophene-3-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.