missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]thiophene-3-acetic acid, 98+%

Product Code. 11327615
Change view
Click to view available options
Quantity:
500 mg
2 g
10 g
Unit Size:
10g
2g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11327615 10 g 10g
11307615 500 mg 500mg
11317615 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11327615 Supplier Thermo Scientific Alfa Aesar Supplier No. L05855.09
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1131-09-5 | C10H8O2S | 192.232 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1131-09-5
Molecular Formula C10H8O2S
Molecular Weight (g/mol) 192.232
MDL Number MFCD00051637
InChI Key VFZQJKXVHYZXMM-UHFFFAOYSA-N
Synonym benzo b thiophene-3-acetic acid, benzo b thiophen-3-yl acetic acid, 3-benzo b thienylacetic acid, acetic acid, 3-benzo b thienyl, 2-benzo b thiophen-3-yl acetic acid, 2-benzo b thiophen-3-ylacetic acid, benzothiophene-3-acetic acid, 2-1-benzothiophen-3-yl acetic acid, 3-benzo b thiopheneacetic acid
PubChem CID 70799
IUPAC Name 2-(1-benzothiophen-3-yl)acetic acid
SMILES C1=CC=C2C(=C1)C(=CS2)CC(=O)O

Specifications

Melting Point 108°C to 112°C
Quantity 10 g
Beilstein 143357
Formula Weight 192.24
Percent Purity ≥98%
Chemical Name or Material Benzo[b]thiophene-3-acetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.