missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzo[b]thiophene-2-sulfonyl chloride, 97%, Thermo Scientific™
Shop All Thermo Scientific Chemicals ProductsDescription
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| CAS | 90001-64-2 |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 96% min. as diethylsulfonamide (HPLC) |
| Packaging | Glass Bottle |
| Molecular Formula | C8H5ClO2S2 |
| Quantity | 1g |
| Synonym | benzo b thiophene-2-sulfonyl chloride, 1-benzothiophene-2-sulfonylchloride, benzo b thiophene-2-sulfonylchloride, benzothiophene-2-sulfonyl chloride, pubchem10221, 2-benzothiophenesulfonyl chloride, benzo b thiophen-2-ylchlorosulfone, 1-benzothiophene-2-sulphonyl chloride, 2-chlorosulphonyl-1-benzothiophene, 1-benzo b thiophene-2-sulfonyl chloride |
| InChI Key | FKIIVBOPAHICHQ-UHFFFAOYSA-N |
| SMILES | C1=CC=C2C(=C1)C=C(S2)S(=O)(=O)Cl |
| IUPAC Name | 1-benzothiophene-2-sulfonyl chloride |
| Show More |
Safety and Handling
Product Identifier
- Benzo¢b!thiophene-2-sulfonyl chloride, 97%
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- EUH029-Contact with water liberates toxic gas.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?