missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]thiophene-2-boronic acid, 97%

Product Code. p-7026688
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11479867

Brand: Thermo Scientific Alfa Aesar B22835.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 98437-23-1 | C8H7BO2S | 178.01 g/mol

Benzo[b]thiophene-2-boronic acid is used in the synthesis of phosphorescent sensor for quantification of copper(II) ion, UV promoted phenanthridine syntheses, Preparation of CYP11B1 inhibitors for treatment of cortisol dependent diseases, PDE4 inhibitors, Suzuki-Miyaura cross-coupling reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzo[b]thiophene-2-boronic acid is used in the synthesis of phosphorescent sensor for quantification of copper(II) ion, UV promoted phenanthridine syntheses, Preparation of CYP11B1 inhibitors for treatment of cortisol dependent diseases, PDE4 inhibitors, Suzuki-Miyaura cross-coupling reactions.

Solubility
Insoluble in water.

Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98437-23-1
Molecular Formula C8H7BO2S
Molecular Weight (g/mol) 178.01
MDL Number MFCD01075674
InChI Key YNCYPMUJDDXIRH-UHFFFAOYSA-N
Synonym 2-benzothienylboronic acid, benzo b thiophen-2-ylboronic acid, benzo b thiophene-2-boronic acid, benzothiophene-2-boronic acid, 1-benzothien-2-ylboronic acid, thianaphthene-2-boronic acid, benzo b thiophene-2-ylboronic acid, 2-benzothiopheneboronic acid, benzothiophen-2-ylboronic acid
PubChem CID 2359
SMILES OB(O)C1=CC2=CC=CC=C2S1

Specifications

Melting Point ∼255°C
Quantity 1 g
Beilstein 1637924
Solubility Information Insoluble in water.
IUPAC Name (1-benzothiophen-2-yl)boronic acid
Formula Weight 178.02
Percent Purity 98%
Chemical Name or Material Benzo[b]thiophene-2-boronic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.