missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]thiophene 1,1-dioxide, 98%

Product Code. 11487986
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11487986 25 g 25g
11477986 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11487986 Supplier Thermo Scientific Alfa Aesar Supplier No. A14063.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 825-44-5 | C8H6O2S | 166.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 825-44-5
Molecular Formula C8H6O2S
Molecular Weight (g/mol) 166.19
MDL Number MFCD00052178
InChI Key FRJNKYGTHPUSJR-UHFFFAOYSA-N
Synonym benzo b thiophene 1,1-dioxide, benzothiophene 1,1-dioxide, thianaphthene 1,1-dioxide, benzo b thiophene, 1,1-dioxide, thianaphthene-1,1-dioxide, benzo b thiophene s,s-dioxide, benzo b thiole-1,1-dione, benzothiophene sulfone, maybridge1_005454
PubChem CID 69997
IUPAC Name 1-benzothiophene 1,1-dioxide
SMILES O=S1(=O)C=CC2=CC=CC=C12

Specifications

Melting Point 137°C to 138°C
Quantity 25 g
Beilstein 116442
Formula Weight 166.2
Percent Purity 98%
Chemical Name or Material Benzo[b]thiophene 1,1-dioxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.