missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]furan, 97+%

Product Code. 11452258
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11452258 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11452258 Supplier Thermo Scientific Alfa Aesar Supplier No. A15225.30
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 271-89-6 | C8H6O | 118.14 g/mol

Benzo[b]furan is used in the manufacture of coumarone-indene resin. It has been used to make floor tiles and other products. It is also used as a coating on grapefruit, lemons, lirnes, oranges, tangelos and tangerines and in the production of COITosion-resistant paints and varnishes, in water-resistant coatings on paper products and fabrics and as adhesives in food containers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzo[b]furan is used in the manufacture of coumarone-indene resin. It has been used to make floor tiles and other products. It is also used as a coating on grapefruit, lemons, lirnes, oranges, tangelos and tangerines and in the production of COITosion-resistant paints and varnishes, in water-resistant coatings on paper products and fabrics and as adhesives in food containers.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Specifications

CAS 271-89-6
Density 1.095
Boiling Point 173°C to 175°C
Flash Point 56°C
Odor Aromatic
Molecular Formula C8H6O
Refractive Index 1.566
MDL Number MFCD00005847
Quantity 250 g
UN Number UN1993
Beilstein 107704
Merck Index 14,1088
Synonym benzofuran, 2,3-benzofuran, coumarone, benzo b furan, cumarone
Solubility Information Not miscible aq. soln..
InChI Key IANQTJSKSUMEQM-UHFFFAOYSA-N
SMILES O1C=CC2=CC=CC=C12
IUPAC Name 1-benzofuran
Molecular Weight (g/mol) 118.14
PubChem CID 9223
ChEBI CHEBI:35260
Formula Weight 118.14
Percent Purity ≥99%
Chemical Name or Material Benzo[b]furan
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.