missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]furan-7-carbaldehyde, Thermo Scientific™

Product Code. 10732947
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10732947 1 g Amber Glass Bottle 1g
10497734 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10732947 Supplier Thermo Scientific Maybridge Supplier No. CC78204DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 95333-14-5
Molecular Formula C9H6O2
Molecular Weight (g/mol) 146.145
MDL Number MFCD08272125
InChI Key RGPUSZZTRKTMNA-UHFFFAOYSA-N
Synonym benzofuran-7-carbaldehyde, 7-benzofurancarboxaldehyde, 7-formylbenzofuran, 7-benzofurancarboxaldehyde 9ci, benzofuran-7-carboxaldehyde, 7-benzo b furancarbaldehyde, benzo b furan-7-carbaldehyde
PubChem CID 11126406
IUPAC Name 1-benzofuran-7-carbaldehyde
SMILES C1=CC2=C(C(=C1)C=O)OC=C2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 146.14
Percent Purity 95%
Chemical Name or Material Benzo[b]furan-7-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.