missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]furan-3-boronic acid, 98%, Thermo Scientific Chemicals

Product Code. 15451579
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 317830-83-4
Molecular Formula C8H7BO3
Molecular Weight (g/mol) 161.95
MDL Number MFCD06801687
InChI Key DFUGYZQSDFQVPU-UHFFFAOYSA-N
Synonym benzofuran-3-ylboronic acid, benzofuran-3-boronic acid, 1-benzofuran-3-yl boronic acid, boronic acid, 3-benzofuranyl, acmc-1csxn, benzo b furan-3-boronic acid, benzofuran-3-yl boronic acid
PubChem CID 23438904
SMILES OB(O)C1=COC2=CC=CC=C12

Specifications

Quantity 1 g
Beilstein 9479377
IUPAC Name 1-benzofuran-3-ylboronic acid
Formula Weight 161.95
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Benzo[b]furan-3-boronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'