missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]furan-3-acetonitrile, 99%

Product Code. 11328946
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11328946 250 mg 250mg
11318946 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11328946 Supplier Thermo Scientific Alfa Aesar Supplier No. L19550.MD
 Restricted Item
Estimated Shipment: 03-12-2026
Añadir a la cesta
Añadir a la cesta
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 52407-43-9 | C10H7NO | 157.172 g/mol

Benzo[b]furan-3-acetonitrile is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzo[b]furan-3-acetonitrile is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Insoluble in water.

Notes
Keep the container tightly sealed. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 52407-43-9
Molecular Formula C10H7NO
Molecular Weight (g/mol) 157.172
MDL Number MFCD02183555
InChI Key BJAJKVZABJZXDC-UHFFFAOYSA-N
Synonym 1-benzofuran-3-ylacetonitrile, 3-benzo b furylacetonitrile, benzofuran-3-acetonitrile, 2-benzofuran-3-yl acetonitrile, benzo b furan-3-acetonitrile, 2-3-benzofuranyl acetonitrile, 2-1-benzofuran-3-yl acetonitrile, 2-benzo b furan-3-ylethanenitrile, pubchem7006, 3-benzofuranylacetonitrile
PubChem CID 2769401
IUPAC Name 2-(1-benzofuran-3-yl)acetonitrile
SMILES C1=CC=C2C(=C1)C(=CO2)CC#N

Especificaciones

Melting Point 38°C to 39°C
Boiling Point 120°C to 122°C (4 mmHg)
Quantity 250 mg
Beilstein 1282785
Solubility Information Insoluble in water.
Formula Weight 157.17
Percent Purity 99%
Chemical Name or Material Benzo[b]furan-3-acetonitrile

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.