missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]furan-2-carboxaldehyde, 97%, Maybridge™

Product Code. 10596684
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10596684 10 g 10g
10493945 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10596684 Supplier Thermo Scientific Maybridge Supplier No. CC06604EA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 4265-16-1
Molecular Formula C9H6O2
Molecular Weight (g/mol) 146.145
InChI Key ADDZHRRCUWNSCS-UHFFFAOYSA-N
Synonym benzo b furan-2-carboxaldehyde, 2-benzofurancarboxaldehyde, benzofuran-2-carbaldehyde, benzofuran-2-carboxaldehyde, 2-formylbenzofuran, 2-benzofurancarbaldehyde, coumarilaldehyde, 1-benzofuran-2-carboxaldehyde, benzofuran-2-aldehyde, benzo b-2-furfural
PubChem CID 61341
IUPAC Name 1-benzofuran-2-carbaldehyde
SMILES C1=CC=C2C(=C1)C=C(O2)C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 10 g
Formula Weight 146.14
Percent Purity 97%
Chemical Name or Material 1-Benzofuran-2-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.