missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]furan-2-carboxaldehyde, 96%

Product Code. p-63879 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10768221

Brand: Thermo Scientific Acros 330140010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4265-16-1 | C9H6O2 | 146.145 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4265-16-1
MDL Number MFCD00015463
InChI Key ADDZHRRCUWNSCS-UHFFFAOYSA-N
Synonym benzo b furan-2-carboxaldehyde, 2-benzofurancarboxaldehyde, benzofuran-2-carbaldehyde, benzofuran-2-carboxaldehyde, 2-formylbenzofuran, 2-benzofurancarbaldehyde, coumarilaldehyde, 1-benzofuran-2-carboxaldehyde, benzofuran-2-aldehyde, benzo b-2-furfural
PubChem CID 61341
IUPAC Name 1-benzofuran-2-carbaldehyde
SMILES C1=CC=C2C(=C1)C=C(O2)C=O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Yellow to Brown
Density 1.2060g/mL
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Molecular Formula C9H6O2
Refractive Index 1.6300 to 1.6330
Quantity 1 g
Specific Gravity 1.206
Solubility Information Solubility in water: .. Other solubilities: acetone,ethanol,halogenated solvents
Molecular Weight (g/mol) 146.14
Formula Weight 146.14
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Benzo[b]furan-2-carboxaldehyde, 96%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.