missing translation for 'onlineSavingsMsg'
Learn More

Benzo-18-crown-6, 97%

Product Code. 10702091 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10702091

Brand: Thermo Scientific Acros 243980010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14098-24-9 | C16H24O6 | 312.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14098-24-9
Molecular Formula C16H24O6
Molecular Weight (g/mol) 312.34
MDL Number MFCD00062741
InChI Key DSFHXKRFDFROER-UHFFFAOYSA-N
Synonym benzo-18-crown-6, b18c6-benzo crown ether, benzo-18-crown 6-ether, 2,3,5,6,8,9,11,12,14,15-decahydrobenzo b 1,4,7,10,13,16 hexaoxacyclooctadecine, 2,3-benzo-1,4,7,10,13,16-hexaoxaoctadec-2-ene, monobenzo-18-crown-6, benzo-18-crown6-ether, 2,3-benzo-18-crown-6, benzo-18-crown, benzo-18-crown 1g
PubChem CID 585779
IUPAC Name 2,5,8,11,14,17-hexaoxabicyclo[16.4.0]docosa-1(22),18,20-triene
SMILES C1COCCOCCOC2=CC=CC=C2OCCOCCO1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 40°C to 45°C
Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 312.34
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Benzo-18-crown-6
Show More Show Less
compliance-icons
Product Identifier
  • Benzo-18-crown-6
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.