missing translation for 'onlineSavingsMsg'
Learn More

Benzo-15-crown-5, 99%, Thermo Scientific™

Product Code. 10665482 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
Packaging:
Glass bottle
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10665482 2.5 g Glass bottle 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10665482 Supplier Thermo Scientific Acros Supplier No. 243990025

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

 

Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY

Specifications

CAS 14098-44-3
CAS Min % 98.5
CAS Max % 100.0
Melting Point 79°C to 82°C
Color Gray to White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C14H20O5
MDL Number MFCD00005945
Quantity 2.5 g
Synonym benzo-15-crown-5, 2,3,5,6,8,9,11,12-octahydrobenzo b 1,4,7,10,13 pentaoxacyclopentadecine, monobenzo-15-crown-5, benzyl 15-crown-5, benzo15c5, denzo15c5, 2,3,5,6,8,9,11,12-octahydro-1,4,7,10,13-benzopentaoxacyclopentadecine, benzo-15-crown-5-ether, benzo-15-crown 5-ether, ccris 3608
Solubility Information Solubility in water: partially soluble
InChI Key FNEPSTUXZLEUCK-UHFFFAOYSA-N
SMILES C1COCCOC2=CC=CC=C2OCCOCCO1
IUPAC Name 2,5,8,11,14-pentaoxabicyclo[13.4.0]nonadeca-1(19),15,17-triene
Molecular Weight (g/mol) 268.31
PubChem CID 84197
ChEBI CHEBI:37444
Formula Weight 268.29
Percent Purity 99%
Physical Form Powder or Flakes
Chemical Name or Material Benzo-15-crown-5, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Benzo-15-crown-5
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.