missing translation for 'onlineSavingsMsg'
Learn More

Benzo-15-crown-5, 98%

Product Code. 11478626
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11478626 5 g 5g
11468626 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11478626 Supplier Thermo Scientific Alfa Aesar Supplier No. A16011.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 14098-44-3 | C14H20O5 | 268.31 g/mol

Benzo-15-crown-5 is used in synthetic chemistry. It plays a major role and involved in complexation through ether oxygen. It is used in phase-transfer catalyst system. It is involved in 'Host-Guest' chemistry to identify the move of essential elements in the body. It can play the part of very complicated biological reactions such as enzyme functions, which finds application in the development of new pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzo-15-crown-5 is used in synthetic chemistry. It plays a major role and involved in complexation through ether oxygen. It is used in phase-transfer catalyst system. It is involved in ′Host-Guest′ chemistry to identify the move of essential elements in the body. It can play the part of very complicated biological reactions such as enzyme functions, which finds application in the development of new pharmaceuticals.

Solubility
Soluble in hot methanol. Slightly soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14098-44-3
Molecular Formula C14H20O5
Molecular Weight (g/mol) 268.31
MDL Number MFCD00005945
InChI Key FNEPSTUXZLEUCK-UHFFFAOYSA-N
Synonym benzo-15-crown-5, 2,3,5,6,8,9,11,12-octahydrobenzo b 1,4,7,10,13 pentaoxacyclopentadecine, monobenzo-15-crown-5, benzyl 15-crown-5, benzo15c5, denzo15c5, 2,3,5,6,8,9,11,12-octahydro-1,4,7,10,13-benzopentaoxacyclopentadecine, benzo-15-crown-5-ether, benzo-15-crown 5-ether, ccris 3608
PubChem CID 84197
ChEBI CHEBI:37444
IUPAC Name 2,5,8,11,14-pentaoxabicyclo[13.4.0]nonadeca-1(19),15,17-triene
SMILES C1COCCOC2=CC=CC=C2OCCOCCO1

Specifications

Melting Point 78°C to 82°C
Quantity 5 g
Beilstein 1624106
Solubility Information Soluble in hot methanol. Slightly soluble in water.
Formula Weight 268.31
Percent Purity 98%
Chemical Name or Material Benzo-15-crown-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.