missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems BenzNuclease DNA and RNA Nuclease (Ultra Pure,Protease Free)

Product Code. 16497129
Click to view available options
Quantity:
10 kU
50 kU
500 kU
Unit Size:
10ku
500ku
50ku

Product Code. 16497129

Brand: ACROBiosystems BEEN3116500KU

Please to purchase this item. Need a web account? Register with us today!

BenzNuclease is a recombinant form of Serratia macescens extracellular endonuclease produced in Escherichia coli cells using a proprietary process at ACRObiosystems. BenzNuclease is a homodimer with monomer molecular masses about 30 kDa. Two disulfide bon

Specifications

Purity >95% as determined by SDS-PAGE
Storage Buffer Tris HCl, pH 8.0, MgCl2, and NaCl
Content And Storage -20°C
Expression System E. coli
Gene Alias DNA and RNA Nuclease,BenzNuclease
Protein Tag Tag Free
Shipping Condition RT
Conjugate Unconjugated
Source BenzNuclease-DNA-RNA-Nuclease (BEE-N3116) is expressed from E.coli cells.
Quantity 500 kU
Product Type BenzNuclease
Form Powder
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.