missing translation for 'onlineSavingsMsg'
Learn More

Benznidazol, MedChemExpress

Product Code. 16468786
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16468786 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16468786 Supplier Medchem Express Supplier No. HYB1548
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Benznidazol (Ro 07-1051) is an antiparasitic medication, with an IC50 of 20.35 μM for Colombian T. cruzi strains, and has been used in the treatment of Chagas disease.

Benznidazol (Ro 07-1051) is an antiparasitic medication, with an IC50 of 20.35 μM for Colombian T. cruzi strains, and has been used in the treatment of Chagas disease.

Specifications

Chemical Name or Material Benznidazol
Physical Form Solid
CAS 22994-85-0
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C12H12N4O3
For Use With (Application) COVID-19-immunoregulation
Synonym Ro 07-1051 Ro 71051
Solubility Information DMSO : 50 mg/mL (192.12 mM; Need ultrasonic)
SMILES O=C(NCC1=CC=CC=C1)CN2C=CN=C2[N+]([O-])=O
Molecular Weight (g/mol) 260.25
Formula Weight 260.25
Percent Purity 99.38%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.