missing translation for 'onlineSavingsMsg'
Learn More

Benzimidazole-5-carboxylic acid, 98%

Product Code. 11304599
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
250 g
Unit Size:
10g
250g
25g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11304599 25 g 25g
11478556 10 g 10g
11488556 50 g 50g
11498556 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11304599 Supplier Thermo Scientific Alfa Aesar Supplier No. A15980.14
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 15788-16-6 | C8H6N2O2 | 162.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15788-16-6
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.15
MDL Number MFCD00011555
InChI Key COYPLDIXZODDDL-UHFFFAOYSA-N
Synonym 1h-benzimidazole-5-carboxylic acid, 5-benzimidazolecarboxylic acid, benzimidazole-5-carboxylic acid, 1h-benzo d imidazole-5-carboxylic acid, 1h-benzimidazole-6-carboxylic acid, 5-carboxybenzimidazole, 1h-1,3-benzodiazole-5-carboxylic acid, 3h-benzo d imidazole-5-carboxylic acid, 1h-benzimidazole-5-carboxylicacid, 1h-1,3-benzodiazole-6-carboxylic acid
PubChem CID 459456
SMILES OC(=O)C1=CC=C2N=CNC2=C1

Specifications

Melting Point >300°C
Quantity 25 g
Beilstein 5686
IUPAC Name 3H-benzimidazole-5-carboxylic acid
Formula Weight 162.15
Percent Purity 98%
Chemical Name or Material Benzimidazole-5-carboxylic acid
compliance-icons
Product Identifier
  • Benzimidazole-5-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 1H-Benzimidazole-6-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.