missing translation for 'onlineSavingsMsg'
Learn More

Benzimidazole-5,6-dicarboxylic acid, 97%

Product Code. 11363676
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11363676 5 g 5g
11353676 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11363676 Supplier Thermo Scientific Alfa Aesar Supplier No. L14754.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 10351-75-4 | C9H6N2O4 | 206.16 g/mol

Benzimidazole-5,6-dicarboxylic acid is a important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzimidazole-5,6-dicarboxylic acid is a important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10351-75-4
Molecular Formula C9H6N2O4
Molecular Weight (g/mol) 206.16
MDL Number MFCD03093058
InChI Key PIPQOFRJDBZPFR-UHFFFAOYSA-N
Synonym benzimidazole-5,6-dicarboxylic acid, 1h-benzo d imidazole-5,6-dicarboxylic acid, 1h-1,3-benzodiazole-5,6-dicarboxylic acid, 1h-benzoimidazole-5,6-dicarboxylic, 1h-benzoimidazole-5,6-dicarboxylic acid, acmc-1bsyc, 5,6-benzimidazoledicarboxylic acid, benzo d imidazole-5,6-dicarboxylic acid
PubChem CID 7023591
IUPAC Name 1H-benzimidazole-5,6-dicarboxylic acid
SMILES OC(=O)C1=C(C=C2N=CNC2=C1)C(O)=O

Specifications

Melting Point ∽295°C (decomposition)
Quantity 5 g
Beilstein 528170
Solubility Information Insoluble in water.
Formula Weight 206.16
Percent Purity 97%
Chemical Name or Material Benzimidazole-5,6-dicarboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.