missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzilic-d10 Acid (rings-d10), CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
Specifications
Specifications
| Chemical Name or Material | Benzilic acid diphenyl-d10 |
| CAS | 1219799-12-8 |
| Molecular Formula | C14 2H10 H2 O3 |
| Quantity | 0.25 g |
| Synonym | Benzilic acid D10 (diphenyl-D10), Benzilic-d10 acid (rings-d10), Benzilic-d10 acid, Di(phenyl-d5)hydroxyacetic acid, 2,2-Bis(pentadeuteriophenyl)-2-hydroxyethanoic acid |
| InChI Formula | InChI=1 S/C14H12O3/c15-13(16)14(17,11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10,17 H,(H,15,16)/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D,9 D,10 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(c([2 H])c1[2 H])C(O)(C(=O)O)c2c([2 H])c([2 H])c([2 H])c([2 H])c2[2 H] |
| IUPAC Name | 2-hydroxy-2,2-bis(2,3,4,5,6-pentadeuteriophenyl)acetic acid |
| Molecular Weight (g/mol) | 238.3 |
| Formula Weight | 238.1414 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction