missing translation for 'onlineSavingsMsg'
Learn More

Benzhydrylamine hydrochloride, polymer-supported, 1% cross-linked, 100-200 mesh, 0.5-1.5 mmol/g on PS-DVB, Thermo Scientific™

Product Code. 11440780
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11440780 25 g 25g
11430780 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11440780 Supplier Thermo Scientific Alfa Aesar Supplier No. 044459.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5267-34-5
Molecular Formula C13H14ClN
Molecular Weight (g/mol) 219.712
MDL Number MFCD03456188
InChI Key CIHWJRSPVJBHGT-UHFFFAOYSA-N
Synonym benzhydrylamine hydrochloride, aminodiphenylmethane hydrochloride, benzhydrylamine, hydrochloride, diphenylmethanamine hydrochloride, benzhydrylammonium chloride, aminodiphenylmethane hcl, 1,1-diphenylmethylamine hydrochloride, diphenylmethylamine, chloride, benzhydrylamin hydrochloride, bha-polystyrene
PubChem CID 78921
IUPAC Name diphenylmethanamine;hydrochloride
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)N.Cl

Specifications

Mesh Size 100 to 200 Mesh
Quantity 25 g
Physical Form Powder
Chemical Name or Material Benzhydrylamine hydrochloride, Polymer-supported, 1% cross-linked, 0.5 to 1.5 mmol/g on PS-DVB
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.