missing translation for 'onlineSavingsMsg'
Learn More

Benzhydrylamine, 97%

Product Code. 11435727
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11435727 100 g 100g
11415727 5 g 5g
11425727 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11435727 Supplier Thermo Scientific Alfa Aesar Supplier No. B24303.22
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 91-00-9 | C13H13N | 183.254 g/mol

Benzhydrylamine is useful for chiral resolution. It acts as an active pharmaceutical ingredient intermediate. It is also employed as a building block for chemical synthesis. Further, it is used in the preparation of furan-2-carboxylic acid benzhydrylamide by reacting with furan-2-carbonyl chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzhydrylamine is useful for chiral resolution. It acts as an active pharmaceutical ingredient intermediate. It is also employed as a building block for chemical synthesis. Further, it is used in the preparation of furan-2-carboxylic acid benzhydrylamide by reacting with furan-2-carbonyl chloride.

Solubility
Miscible with chloroform, dimethylsulfoxide and methanol. Slightly miscible with water.

Notes
Air sensitive and moisture sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong acids and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-00-9
Molecular Formula C13H13N
Molecular Weight (g/mol) 183.254
MDL Number MFCD00008059
InChI Key MGHPNCMVUAKAIE-UHFFFAOYSA-N
Synonym aminodiphenylmethane, benzhydrylamine, 1,1-diphenylmethylamine, diphenylmethyl amine, 1,1-diphenylmethanamine, alpha-aminodiphenylmethane, methanamine, 1,1-diphenyl, alpha-phenylbenzylamine, benzhydrylamin, benzhydryl amine
PubChem CID 7036
IUPAC Name diphenylmethanamine
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)N

Specifications

Melting Point 11°C to 13°C
Density 1.064
Boiling Point 294°C to 296°C
Flash Point >110°C (230°F)
Linear Formula (C6H5)2CHNH2
Refractive Index 1.595
Quantity 100 g
UN Number UN2735
Beilstein 776434
Sensitivity Air Sensitive
Merck Index 14,1076
Solubility Information Miscible with chloroform,dimethylsulfoxide and methanol; Slightly miscible with water.
Formula Weight 183.25
Percent Purity 97%
Chemical Name or Material Benzhydrylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.