missing translation for 'onlineSavingsMsg'
Learn More

Benzhydryl chloride, 98%

Product Code. 11426486
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11426486 25 g 25g
11436486 100 g 100g
11446486 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11426486 Supplier Thermo Scientific Alfa Aesar Supplier No. B24764.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 90-99-3 | C13H11Cl | 202.681 g/mol

Benzhydryl chloride was used as initiator during the controlled radical polymerization of styrene catalyzed by ionic iron complex. It was also used as the starting reagent in the synthesis of trimethylhydroquinone derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzhydryl chloride was used as initiator during the controlled radical polymerization of styrene catalyzed by ionic iron complex. It was also used as the starting reagent in the synthesis of trimethylhydroquinone derivatives.

Solubility
Insoluble in water.

Notes
Store in a tightly sealed container. Store away from oxidizing agents, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-99-3
Molecular Formula C13H11Cl
Molecular Weight (g/mol) 202.681
MDL Number MFCD00000855
InChI Key ZDVDCDLBOLSVGM-UHFFFAOYSA-N
Synonym benzhydryl chloride, chlorodiphenylmethane, chloromethylene dibenzene, diphenylchloromethane, diphenylmethyl chloride, benzene, 1,1'-chloromethylene bis, chloro phenyl methyl benzene, methane, chlorodiphenyl, unii-cn9n9ayv4b, 1,1'-chloromethylene bisbenzene
PubChem CID 7035
IUPAC Name [chloro(phenyl)methyl]benzene
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)Cl

Specifications

Melting Point 17°C to 20°C
Density 1.14
Boiling Point 139°C to 140°C (3 mmHg)
Flash Point 160°C (320°F)
Odor Pungent
Refractive Index 1.595
Quantity 25 g
UN Number UN3265
Beilstein 607925
Solubility Information Insoluble in water.
Formula Weight 202.69
Percent Purity 98%
Chemical Name or Material Benzhydryl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.