missing translation for 'onlineSavingsMsg'
Learn More

Benzhydryl Bromide, TRC

Product Code. 30344406
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30344406 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30344406 Supplier Toronto Research Chemicals Supplier No. TRCB1950005G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzhydryl Bromide
CAS 776-74-9
Molecular Formula C13H11Br
Quantity 5 g
Synonym 1,1'-(Bromomethylene)bisbenzene, Bromomethylene)dibenzene, 1,1'-(Bromomethylene)bis[benzene], 1-[Bromo(phenyl)methyl]benzene, Bromodiphenylmethane, Diphenylbromomethane, Diphenylmethyl Bromide, NSC 39226, alpha-Bromodiphenylmethane, alpha-Phenylbenzyl Bromide
InChI Formula InChI=1S/C13H11Br/c14-13(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10,13H
SMILES BrC(c1ccccc1)c2ccccc2
IUPAC Name [bromo(phenyl)methyl]benzene
Molecular Weight (g/mol) 247.13
Formula Weight 246.0044
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.