missing translation for 'onlineSavingsMsg'
Learn More

Benzhydrol, TRC

Product Code. 17954155
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17954155 5 g 5g
17943725 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17954155 Supplier Toronto Research Chemicals Supplier No. TRCB1938005G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-01-0
Molecular Formula C13 H12 O
Molecular Weight (g/mol) 184.23
Synonym Diphenylmethanol, Benzhydrol, Cyclizine Hydrochloride Imp. B (EP), Dimenhydrinate Imp. I (EP), Diphenhydramine Hydrochloride Imp. D (EP), Ebastine Imp. A (EP), Benzenemethanol, α-phenyl- (9CI, ACI), Benzhydrol (8CI), α-Phenylbenzenemethanol (ACI), 1,1-Diphenylmethanol, Benzhydryl alcohol, Benzohydrol, Diphenylcarbinol, Diphenylmethyl alcohol, Hydroxydiphenylmethane, NSC 32150, α-Phenylbenzyl alcohol
IUPAC Name diphenylmethanol
SMILES OC(c1ccccc1)c2ccccc2

Specifications

Quantity 5 g
InChI Formula InChI=1S/C13H12O/c14-13(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10,13-14H
Formula Weight 184.0888
Percent Purity >95%
Chemical Name or Material Diphenylmethanol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.