missing translation for 'onlineSavingsMsg'
Learn More

Benzethonium chloride, MedChemExpress

Product Code. 16498526
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498526 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16498526 Supplier Medchem Express Supplier No. HYB0942
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Benzethonium chloride inhibit human recombinant α7 and α4β2 neuronal nicotinic acetylcholine receptors in Xenopus oocytes.

Benzethonium chloride inhibit human recombinant α7 and α4β2 neuronal nicotinic acetylcholine receptors in Xenopus oocytes.

Specifications

Chemical Name or Material Benzethonium chloride
CAS 121-54-0
Color White
Purity Grade Notes Research
Molecular Formula C27H42ClNO2
Quantity 100 mg
Solubility Information DMSO : 33.33 mg/mL (74.38 mM; Need ultrasonic) ∣H2O : 20 mg/mL (44.63 mM; Need ultrasonic)
SMILES CC(C)(C)CC(C1=CC=C(OCCOCC[N+](C)(C)CC2=CC=CC=C2)C=C1)(C)C.[Cl-]
Molecular Weight (g/mol) 448.08
Formula Weight 448.08
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.