missing translation for 'onlineSavingsMsg'
Learn More

Benzethonium Chloride 0.004 M According to European Pharmacopoeia (EP), Reagecon
BIOPHARMA

Product Code. 11526743 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11526743 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11526743 Supplier Reagecon Supplier No. HY0041

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Fully factorized, high-purity, stable product developed and tested for titrations.

  • European Pharmacopoeia (EP) Reference Standard, European Pharmacopoeia (EP) Reference Material
  • Accurate, stable, reproducible and fully factorized.
  • Titrations performed with this product, exhibit defined stoichiometry, are quantitative, establish an equilibrium that is definitive and fast and provide unambiguous results.
  • Stability is proven (either opened or unopened) for the shelf life of this product.
  • Certified to a specification of ± 0.2%.
  • Product is a reagent of known concentration, that is dispensed from a burette or other dispensing apparatus to a sample (analyte), until a reaction between the two liquids is judged to be complete.
  • May be used for manual titrations, or automatic titrations, to determine the end points by detection as detailed in several EP monographs.

Specifications

Description Accuracy ± 0.2%
Packaging HDPE Bottle GL45
Physical Form Liquid
Technique Titration, EP Monographs
Quantity 1 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.