missing translation for 'onlineSavingsMsg'
Learn More

Benzenesulfonyl Hydrazide, 98%, ACROS Organics™

Product Code. 10356260 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
100g; Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10356260 100g; Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10356260 Supplier Thermo Scientific Acros Supplier No. 105301000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 80-17-1
CAS Min % 97.5
CAS Max % 100.0
Melting Point 100°C to 104°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C6H8N2O2S
Linear Formula C6H5SO2NHNH2
MDL Number MFCD00007583,MFCD08064450
Quantity 100 g
Beilstein 11, 52
Fieser 07,13
Merck Index 15, 1074
Synonym benzenesulfonyl hydrazide, porofor, benzenesulfonic acid, hydrazide, porofor bsh, benzenesulfohydrazide, phenylsulfonylhydrazine, genitron bsh, celogen bsh, benzenesulfonylhydrazine, phenylsulfohydrazide
InChI Key VJRITMATACIYAF-UHFFFAOYSA-N
SMILES NNS(=O)(=O)C1=CC=CC=C1
IUPAC Name benzenesulfonohydrazide
Molecular Weight (g/mol) 172.20
PubChem CID 65723
Formula Weight 172.2
Percent Purity ≥97.5%
Physical Form Powder
Chemical Name or Material Benzenesulfonyl hydrazide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.