missing translation for 'onlineSavingsMsg'
Learn More

Benzenesulfonyl Chloride, TRC

Product Code. 30342633
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
Product Code. Quantity unitSize
30342633 25 g 25g
1 options
This item is not returnable. View return policy

Product Code. 30342633

Brand: Toronto Research Chemicals TRCB19000025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzenesulfonyl Chloride
CAS 98-09-9
Molecular Formula C6H5ClO2S
Quantity 25 g
Synonym Benzene sulfochloride, Benzenesulfonic chloride, Benzenesulphonyl chloride, NSC 2864, Phenylsulfonyl chloride
InChI Formula InChI=1S/C6H5ClO2S/c7-10(8,9)6-4-2-1-3-5-6/h1-5H
SMILES ClS(=O)(=O)c1ccccc1
IUPAC Name benzenesulfonyl chloride
Molecular Weight (g/mol) 176.92
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.