missing translation for 'onlineSavingsMsg'
Learn More

Benzenesulfonyl chloride, 98%

Product Code. 11459822
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11459822 5000 g 5000g
11449822 250 g 250g
11439822 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11459822 Supplier Thermo Scientific Alfa Aesar Supplier No. A10849.0I
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 98-09-9 | C6H5ClO2S | 176.614 g/mol

Benzenesulfonyl chloride reacts with Grignard reagents to form oxindoles from N-unsubstituted indoles. It is widely used to check the assay of thiamine in different food products. It is involved in the synthesis of alpha-disulfones, sulfonamides and sulfoante esters as precursor. It is a derivatization reagent for the determination of various amines in waste water and surface water at the sub-ppb level by gas chromatography-mass spectrometry. It is common reagent used in Hinsberg test for detection and distinguishing the type of amines as primary, secondary and tertiary amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzenesulfonyl chloride reacts with Grignard reagents to form oxindoles from N-unsubstituted indoles. It is widely used to check the assay of thiamine in different food products. It is involved in the synthesis of alpha-disulfones, sulfonamides and sulfoante esters as precursor. It is a derivatization reagent for the determination of various amines in waste water and surface water at the sub-ppb level by gas chromatography-mass spectrometry. It is common reagent used in Hinsberg test for detection and distinguishing the type of amines as primary, secondary and tertiary amines.

Solubility
Insoluble in water. Soluble in ether and alcohol.

Notes
Moisture sensitive. Incompatible with water, strong oxidizing agents, strong bases, methyl formamide and dimethyl sulfoxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-09-9
Molecular Formula C6H5ClO2S
Molecular Weight (g/mol) 176.614
MDL Number MFCD00007426
InChI Key CSKNSYBAZOQPLR-UHFFFAOYSA-N
Synonym phenylsulfonyl chloride, benzene sulfochloride, benzenesulfonic chloride, benzenosulfochloride, benzene sulfonechloride, benzenosulfochlorek, benzenesulfonic acid chloride, benzenesulphonyl chloride, benzolsulfochloride, benzenesulfochloride
PubChem CID 7369
IUPAC Name benzenesulfonyl chloride
SMILES C1=CC=C(C=C1)S(=O)(=O)Cl

Specifications

Melting Point 14°C to 15°C
Density 1.377
Boiling Point 252°C
Flash Point 132°C (270°F)
Odor Irritating
Refractive Index 1.5525
Quantity 5000 g
UN Number UN2225
Beilstein 606926
Sensitivity Moisture sensitive
Merck Index 14,1072
Solubility Information Insoluble in water. Soluble in ether and alcohol.
Formula Weight 176.62
Percent Purity 98%
Chemical Name or Material Benzenesulfonyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.