missing translation for 'onlineSavingsMsg'
Learn More

Benzenesulfonic acid, 94%

Product Code. 15493117
Change view
Click to view available options
Quantity:
500 g
2500 g
Unit Size:
2500g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15493117 500 g 500g
15483117 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15493117 Supplier Thermo Scientific Alfa Aesar Supplier No. A16148.36
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 98-11-3 | C6H6O3S | 158.171 g/mol

Benzenesulfonic acid is mainly consumed by conversion to other specialty chemicals. A variety of pharmaceutical drugs are prepared as salts of benzenesulfonic acid and are known as besylates or besilates. The alkali metal salt of benzenesulfonic acid was once widely used in the production of phenol. It acts as an acid catalyst for direct esterification of amino acids and peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzenesulfonic acid is mainly consumed by conversion to other specialty chemicals. A variety of pharmaceutical drugs are prepared as salts of benzenesulfonic acid and are known as besylates or besilates. The alkali metal salt of benzenesulfonic acid was once widely used in the production of phenol. It acts as an acid catalyst for direct esterification of amino acids and peptides.

Solubility
Soluble in water and alcohol. Slightly soluble in benzene. Insoluble in non-polar solvents.

Notes
It is hygroscopic. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-11-3
Molecular Formula C6H6O3S
Molecular Weight (g/mol) 158.171
MDL Number MFCD00011689
InChI Key SRSXLGNVWSONIS-UHFFFAOYSA-N
Synonym benzenesulphonic acid, phenylsulfonic acid, besylic acid, benzene sulphonic acid, benzenemonosulfonic acid, benzene sulfonic acid, kyselina benzensulfonova, benzensulfonic acid, ccris 4595, kyselina benzensulfonova czech
PubChem CID 7371
ChEBI CHEBI:64455
IUPAC Name benzenesulfonic acid
SMILES C1=CC=C(C=C1)S(=O)(=O)O

Specifications

Melting Point ∼45°C to 50°C
Density 1.32
Flash Point >110°C (230°F)
Quantity 500 g
UN Number UN2585
Beilstein 742513
Sensitivity Hygroscopic
Merck Index 14,1070
Solubility Information Soluble in water and alcohol. Slightly soluble in benzene. Insoluble in non-polar solvents.
Formula Weight 158.18
Percent Purity 94%
Chemical Name or Material Benzenesulfonic acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Benzenesulfonic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Benzenesulfonic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.