missing translation for 'onlineSavingsMsg'
Learn More

Benzenesulfonamide, 98+%

Product Code. 11404253
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11404253 5000 g 5000g
11494243 250 g 250g
11484243 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11404253 Supplier Thermo Scientific Alfa Aesar Supplier No. A10891.0I
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 98-10-2 | C6H7NO2S | 157.187 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
The presence of the benzenesulfonylamido (BS) groups shifted the activity of classic antimicrobial fluoroquinolones from being more active against Gram-negative to Gram-positive strains. Biospecific adsorption of carbonic anhydrase to self-assembled monolayers of alkanethiolates that present benzenesulfonamide groups on gold. Biospecific binding of carbonic anhydrase to mixed sams presenting benzenesulfonamide ligands led to a model system for studying lateral steric effects. In engineering the specificity of antibacterial fluoroquinolones, benzenesulfonamide modifications at c-7 of ciprofloxacin change its primary target instreptococcus pneumoniae from topoisomerase iv to gyrase. Polar substitutions in the benzenesulfonamide ring of celecoxib afford a potent 1,5-diarylpyrazole class of COX-2 inhibitors.

Solubility
Soluble in methanol (25 mg/ml), and water (4.3 mg/ml at 16°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-10-2
Molecular Formula C6H7NO2S
Molecular Weight (g/mol) 157.187
MDL Number MFCD00007930
InChI Key KHBQMWCZKVMBLN-UHFFFAOYSA-N
Synonym benzenesulphonamide, benzosulfonamide, benzolsulfonamide, phenyl sulfonamide, benzene sulfonamide, phenylsulfonamide, m and b 7973, chembl27601, benzolsulfonamid, benzensulfonamide
PubChem CID 7370
IUPAC Name benzenesulfonamide
SMILES C1=CC=C(C=C1)S(=O)(=O)N

Specifications

Melting Point 151°C to 154°C
Flash Point 250°C (482°F)
Odor Odorless
Quantity 5000 g
Beilstein 1100566
Solubility Information Soluble in methanol (25mg/ml),and water (4.3mg/ml at 16°C).
Formula Weight 157.19
Percent Purity ≥98%
Chemical Name or Material Benzenesulfonamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.