missing translation for 'onlineSavingsMsg'
Learn More

Benzenesulfinic Acid Sodium Salt, TRC

Product Code. 30347640
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30347640 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30347640 Supplier Toronto Research Chemicals Supplier No. TRCB12060025G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzenesulfinic Acid Sodium Salt
CAS 873-55-2
Molecular Formula C6H5NaO2S
Quantity 25 g
Synonym NSC 135147, NSC 30629, Sodium Benzenesulfinate, Sodium Phenylsulfinate
InChI Formula InChI=1S/C6H6O2S.Na/c7-9(8)6-4-2-1-3-5-6;/h1-5H,(H,7,8);/q;+1/p-1
SMILES [Na+].[O-]S(=O)c1ccccc1
IUPAC Name sodium;benzenesulfinate
Molecular Weight (g/mol) 164.16
Formula Weight 163.9908
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.